Learn More
Medchemexpress LLC Methyl tetracosanoate | 2442-49-1 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN843850MG
Encompass_Preferred
Methyl tetracosanoate is a fatty acid methyl ester with anti-diabetic activity. It increases glucose uptake on differentiated 3T3-L1 adipocytes and elevates GLUT4 mRNA expression.
  • Anti-diabetic activity
  • Increases glucose uptake on differentiated 3T3-L1 adipocytes
  • Elevates GLUT4 mRNA expression
Catalog No. 50-004-17280