Learn More
Medchemexpress LLC Methyl tetradecanoate-d27 | 434349-93-6 | 269.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW004288S
Encompass_Preferred
Methyl tetradecanoate-d27 is a deuterium labeled methyl myristate, a saturated fatty acid methyl ester derived from myristic acid. It demonstrates high melanin induction in B16F10 melanoma. Deuteration is gaining attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements for quantitation during drug development.
Catalog No. 50-004-47742