Learn More
Medchemexpress LLC Methyl vanillate | 3943-74-6 | 99.88% | C9H10O4 | 100 G
SDP
Supplier:  Medchemexpress LLC HY75342100G
Encompass_Preferred
Methyl vanillate, found in Oryza sativa Linn., acts as a Wnt/β-catenin pathway activator. This benzoate ester, a methyl ester of vanillic acid, also functions as an antioxidant and a plant metabolite.
  • Acts as a Wnt/β-catenin pathway activator
  • Functions as an antioxidant
  • Functions as a plant metabolite
  • Increases expression and nuclear translocation of β-catenin in calvarial osteoblasts
  • Elevates ALP activity in calvarial osteoblasts
  • Solid form storage: 4°C under nitrogen
  • Solution storage: -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-004-33832