Learn More
Medchemexpress LLC Methyl vanillate | 3943-74-6 | 99.88% | C9H10O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY75342500MG
Encompass_Preferred
Methyl vanillate is an ingredient found in *Oryza sativa Linn.*, and acts as a Wnt/β-catenin pathway activator. It is a benzoate ester, specifically the methyl ester of vanillic acid, and functions as an antioxidant and a plant metabolite.
  • Acts as a Wnt/β-catenin pathway activator
  • Possesses antioxidant properties
  • Increases expression and nuclear translocation of β-catenin in calvarial osteoblasts
  • Dose-dependently elevates ALP activity in calvarial osteoblasts
Catalog No. 50-004-33674