Learn More
Cayman Chemical methyl1 5fluoropentyl1H 10mg
SDP
Supplier:  Cayman Chemical 1467510 MG
Encompass_Preferred
An analog of 5-fluoro PB-22 that lacks the quinoline group; intended for forensic and research applications
Catalog No. 50-296-9533