Learn More
Cayman Chemical -MethylamIno-L-AlnInhydr 5mg
SDP
Supplier:  Cayman Chemical 232875 MG
Encompass_Preferred
A neurotoxic non-proteinogenic amino acid; cytotoxic to mouse primary cortical neurons at 3 mM; stimulates release of glutamate and induces oxidative stress in the same cells at 3 mM; enhances amyloid-β (25-35)-induced neuronal cell death in mouse primary cortical neurons at 0.1 mM; induces wet-dog shakes, rigidity, and clonic convulsions in rats at 500 UG/animal; induces parkinsonism in cynomolgus monkeys; has been found in commercial seafood
Catalog No. 50-299-3740