Learn More
Medchemexpress LLC Methylamino-PEG1-Boc | 1807521-03-4 | 95.0% | 203.28 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140154500MG
Encompass_Preferred
Methylamino-PEG1-Boc is a PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs, which are essential tools in targeted protein degradation research.
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49801