Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylboronic acid-d3 | 1332481-37-4 | MFCD12761137 | 99.8% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY34449S10MG

Encompass_Preferred

Methylboronic acid-d3 is the deuterium-labeled form of methylboronic acid supplied as a white to off-white solid reagent for research use. It is characterized by CAS 1332481-37-4, molecular formula CH2D3BO2, molecular weight 62.88 g/mol, and high purity suitable for analytical and synthetic applications.

  • Deuterium-labeled methylboronic acid (methyl-D3)
  • CAS 1332481-37-4
  • Molecular formula CH2D3BO2
  • Molecular weight 62.88 g/mol
  • Purity 99.8%
  • White to off-white solid (powder)
  • Storage: powder at -20°C; in solvent -80°C (6 months) or -20°C (1 month)
  • For research use as a synthetic building block or isotope-labeled standard

Catalog No. 50-002-97620


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.