Learn More
Cayman Chemical Methylcarbamyl PAF C-8 5mg
SDP
Supplier:  Cayman Chemical 90003325 MG
Encompass_Preferred
A C-8 analog of methylcarbamyl PAF C-16, a stable analog of PAF C-16 with a half-life greater than 100 minutes in platelet poor plasma; induces G1-phase cell cycle arrest, suggesting a potential role in the inhibition of oncogenic transformation
Catalog No. 50-289-3985