Learn More
Cayman Chemical Methylchloroisothiazolinone/Methylisothiazolinone Mixture, 5ml
SDP
Supplier:  Cayman Chemical 274705
Encompass_Preferred
A mixture of isothiazolinone biocides; effective against Gram-positive and Gram-negative bacteria (MICs = 0.0002, 0.0002, 0.00005, and 0.00005% (w/w) for S. aureus, P. aeruginosa, A. niger, and C. albicans, respectively); can elicit contact sensitization
Catalog No. NC1550420