Learn More
Selleck Chemical LLC Methylcobalamin S4994-1g
SDP
Supplier:  Selleck Chemical LLC S49941G
Encompass_Preferred
Methylcobalamin (Mecobalamin Vancomin Methycobal Cobametin) is one active form of vitamin B12 which can directly participate in homocysteine metabolism It is used to treat some nutritional diseases and other diseases in clinic such as Alzheimer s disease and rheumatoid arthritis
Catalog No. 50-311-2063