Learn More
Medchemexpress LLC Methylcobalamin (standard) | 13422-55-4 | 96.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0586R100MG
Encompass_Preferred
Methylcobalamin (Standard) is an analytical standard, also known as Mecobalamin (Standard) or CH3-B12 (Standard). It serves as a reference standard for assay and is intended for research and analytical applications. It is commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Used as a reference standard for assay
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
  • Intended for research and analytical applications only
Catalog No. 50-004-08335