Learn More
Cayman Chemical MethylcyclodextrIn 10g
SDP
Supplier:  Cayman Chemical 2163310 G
Encompass_Preferred
A randomly methylated form of β-cyclodextrin; has been used to improve the aqueous solubility of various compounds; has been used to extract cholesterol from lipid membranes; reduces α-synuclein levels in the membrane and detergent-insoluble fractions from B103 neuroblastoma cells transfected with human α-synuclein at 5 mM; reduces α-synuclein levels in mouse brain in a transgenic model of α-synucleinopathy
Catalog No. 50-298-6286