Learn More
Strem, An Ascensus Company (MeCp)(COD)Ir(I), 3N 1g
SDP
Supplier:  Strem, An Ascensus Company 7750001G
Encompass
CAS# 132644-88-3. 1g. (Methylcyclopentadienyl)(1,5-cyclooctadiene)iridium(I), 99% (99.9%-Ir). MFCD01631287. Molecular Weight: 379.53. Molecular Formula: (C6H7)(C8H12)Ir. Color/form: white to off-white pwdr. Strem# 77-5000. http://www.strem.com/catalog/v/77-5000/
Catalog No. 50-901-13150