Learn More
Selleck Chemical LLC Methyldopa 50mg 555-30-6
SDP
Supplier:  Selleck Chemical LLC S164250MG
Encompass_Preferred
Methyldopa (Aldomet) is a DOPA decarboxylase competitive inhibitor with an ED50 of 21.8 mg/kg. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8359