Learn More
Medchemexpress LLC Methylisothiazolinone | 2682-20-4 | MFCD01742315 | 99.5% | 115.15 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW010520
Methylisothiazolinone is a bacterial and fungal inhibitor and preservative, as well as a sensitizer. It can activate matrix metalloproteinases (MMPs) in human bronchial epithelial cells to induce apoptosis and inflammatory response. Methylisothiazolinone can also promote the development of atopic dermatitis in mice by disrupting Th2/Th17 related immune responses and cause mitochondrial damage in the endothelium of rat cerebral blood vessels.
- Bacterial and fungal inhibitor
- Preservative agent
- Activates matrix metalloproteinases (MMPs)
- Promotes atopic dermatitis development in mice
- Causes mitochondrial damage in rat cerebral blood vessels endothelium
- Solid appearance, white to off-white color
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.