Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylmalonic acid-d3 | 42522-59-8 | MFCD00144783 | 98.9% | 121.11 g/mol | C4H3D3O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY103395S5MG

Encompass_Preferred

Deuterium-labeled methylmalonic acid (methylpropanedioic acid-d3) supplied as a solid stable isotope standard for analytical and research applications. It is commonly used as an internal standard for mass spectrometry and tracer studies, and is relevant to metabolic and clinical research involving vitamin B12 assessment.

  • Deuterium (d3) labeled analog of methylmalonic acid.
  • High purity (98.9%).
  • Molecular weight 121.11 g/mol.
  • Chemical formula C4H3D3O4.
  • Supplied as a 5 mg solid.
  • Powder storage: -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-44682


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.