Learn More
Medchemexpress LLC Methylmalonyl-CoA tetralithium | 104809-02-1 | 98% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY126791A1MG
Encompass_Preferred
Methylmalonyl-CoA tetralithium is a catabolite of valine, isoleucine, methionine, threonine, odd-chain fatty acids, and cholesterol. It is converted to succinyl-CoA by enzymatic reaction of methylmalonyl-CoA mutase (MCM) with coenzyme vitamin B12.
  • Endogenous metabolite
  • Converted to succinyl-CoA by enzymatic reaction of methylmalonyl-CoA mutase (MCM) with coenzyme vitamin B12.
Catalog No. 50-003-38881