Learn More
Cayman Chemical MethylnaltrexonbromIde 10mg
SDP
Supplier:  Cayman Chemical 2407210 MG
Encompass_Preferred
A peripherally restricted µ-opioid receptor antagonist (Ki = 10 nM; IC50s = 130.7 and 0.43 nM in rat brain membranes prepared with sodium chloride or water, respectively); selective for µ-opioid receptors over δ- and κ-opioid receptors (Kis = 630.95 and 31.62 nM, respectively); decreases spontaneous mechanical activity in ileum isolated from morphine-treated guinea pigs when administered at doses of 1 and 10 µM; does not inhibit heroin self-administration in rats; does not induce withdrawal syndrome in morphine-dependent rhesus monkeys when administered at doses up to 32 mg/kg
Catalog No. 50-293-2838