Learn More
Medchemexpress LLC Methylnissolin-3-O-glucoside | 94367-42-7 | 20 MG

Supplier: Medchemexpress LLC HYN247320MG
Methylnissolin-3-O-glucoside, also known as Methylnissolin-3-O-β-D-glucoside, is a flavonoid with anti-inflammatory effects. Isolated from the roots of Astragalus membranaceus, this compound is supplied for research purposes.
- Purity of 99.91%.
- Molecular weight: 462.45.
- Chemical formula: C23H26O10.
- Appearance: White to off-white solid.
- Structure classification: Flavonoids.
- Initial source: Plants, Leguminosae, Astragalus membranaceus var. mongholicus.
- Storage recommendations: 4°C, protect from light; in solvent at -80°C for 6 months or -20°C for 1 month, protected from light.
- Solubility in DMSO: 100 mg/mL (216.24 mM).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.