Learn More
Medchemexpress LLC Methylnissolin | 73340-41-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN24845MG
Encompass_Preferred
Methylnissolin (Astrapterocarpan) is an osteoclast inhibitor with anti-inflammatory, neuroprotective, and antioxidant activities. It modulates the MAPK and PI3K/AKT pathways, inhibiting phosphorylation of MAPK1, AKT1, and PDGF-BB-induced ERK1/2. This compound is valuable for research in areas such as ischemic stroke, osteoporosis, cardiovascular diseases, and skin aging.
  • Acts as an osteoclast inhibitor
  • Displays anti-inflammatory, neuroprotective, and antioxidant properties
  • Modulates MAPK and PI3K/AKT signaling pathways
  • Reduces proinflammatory mediators and ROS levels
  • Increases antioxidant enzymes and decreases osteoclastogenesis markers
  • Research applications include ischemic stroke, osteoporosis, cardiovascular diseases, and skin aging
Catalog No. 50-004-20605