Learn More
Medchemexpress LLC Methylnissolin (astrapterocarpan) | 73340-41-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN24841MG
Encompass_Preferred
Methylnissolin (Astrapterocarpan) is an osteoclast inhibitor exhibiting anti-inflammatory, neuroprotective, and antioxidant activities. It is applicable to research related to ischemic stroke, osteoporosis, cardiovascular diseases, and skin aging.
  • Downregulates the activation of MAPK and PI3K/AKT pathways
  • Inhibits the phosphorylation of MAPK1 and AKT1
  • Blocks PDGF-BB-induced phosphorylation of ERK1/2
  • Reduces the expression and secretion of proinflammatory mediators
  • Decreases intracellular ROS levels
  • Upregulates antioxidant enzymes
  • Downregulates osteoclastogenesis markers
Catalog No. 50-004-19996