Learn More
ATCC Methylorubrum extorquens
Supplier:  ATCC 14718
Encompass
Methylorubrum extorquens strain NCIB 9133 [D 355; AM1, R. Hugh 1924] is a whole-genome sequenced bacterial strain that produces glutamate dehydrogenase.
Catalog No. 50-238-2753