Learn More
Cayman Chemical MethylpIperIdIno pyrazole 10mg
SDP
Supplier:  Cayman Chemical 1386310 MG
Encompass_Preferred
A highly selective ERα receptor antagonist (Ki = 5.6 nM; IC50 = 80 nM)
Catalog No. 50-295-5195