Learn More
Selleck Chemical LLC Methylprednisolone S1733-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S173310MM/1ML
Encompass_Preferred
Methylprednisolone is a synthetic glucocorticoid receptor agonist used to achieve prompt suppression of inflammation Methylprednisolone activates ACE2 and reduces IL-6 levels thus improves severe or critical COVID-19 Methylprednisolone markedly reduces autophagy and apoptosis
Catalog No. 50-136-1577