Learn More
Medchemexpress LLC Methylprednisolone | 83-43-2 | 99.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0260500MG
Encompass_Preferred
Methylprednisolone is a synthetic corticosteroid known for its anti-inflammatory and immunomodulating properties. It can improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. It is also used in studies of bronchial inflammation, acute bronchitis, arthritis, various respiratory diseases, and may improve neurological recovery in acute spinal cord injury.
  • Synthetic corticosteroid with anti-inflammatory and immunomodulating properties
  • Improves severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels
  • Commonly used for anti-inflammatory purposes
  • May improve neurological recovery in acute spinal cord injury
  • Targets Glucocorticoid Receptor, Autophagy, SARS-CoV, and Bacterial pathways
  • Relevant to research in cardiovascular disease, infection, neurological disease, metabolic disease, inflammation/immunology, and cancer
Catalog No. 50-004-06151