Learn More
Medchemexpress LLC Methylprednisolone | 83-43-2 | MFCD00010591 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0260R50MG
Encompass_Preferred
Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. It is noted to improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. This product is for research use only.
  • Synthetic corticosteroid
  • Anti-inflammatory properties
  • Immunomodulating properties
  • Activates ACE2
  • Reduces IL-6 levels
  • For research use only
Catalog No. 50-004-05830