Learn More
Medchemexpress LLC Methylprednisolone aceponate | 86401-95-8 | MFCD00866150 | 98.9% | 472.57 g/mol | C27H36O7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10302550MG
Encompass_Preferred
Methylprednisolone aceponate is an analytical/reference standard of the corticosteroid methylprednisolone aceponate intended for research and analytical applications. Supplied in a small 50 mg quantity, this standard is used for chromatographic method development, calibration, and quality control.
  • Analytical reference standard for research use.
  • High purity: 98.9% (LCMS, lot-specific COA available).
  • Small 50 mg pack suitable for method development and calibration.
  • Molecular formula C27H36O7, molecular weight 472.57 g/mol.
  • Suitable for chromatography and instrumental analysis.
Catalog No. 50-002-46988