Learn More
Medchemexpress LLC Methylprednisolone acetate | 53-36-1 | 95.1% | 416.51 | 50 MG

Supplier: Medchemexpress LLC HY1368150MG
Methylprednisolone acetate is a corticosteroid hormone derived from prednisolone. It is known for its ability to relieve pain and reduce swelling associated with arthritis and other joint disorders in living organisms.
- Corticosteroid hormone: Acts as a prednisolone derivative.
- Pain relief: Effective in alleviating discomfort.
- Reduces swelling: Helps mitigate inflammation in joint disorders.
- Research use only: Primarily intended for scientific studies.
- High purity: Available with a purity of 95.06%.
- Variety of solvents: Soluble in DMSO for in vitro applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.