Learn More
Medchemexpress LLC Methylprednisolone acetate | 53-36-1 | MFCD00200356 | 95.1% | 416.51 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1368110MM/1ML
Encompass_Preferred
Methylprednisolone acetate is a prednisolone derivative and a corticosteroid hormone. It is known to relieve pain and swelling associated with arthritis and other joint disorders in vivo. This product appears as a white to off-white solid.
  • Corticosteroid hormone
  • Relieves pain and swelling associated with arthritis
  • Used in joint disorder research
  • Targeted disease areas include neurological, eye or ear disease, inflammation or immune system disease, and pain
  • Involves signaling pathways such as Immunology/Inflammation, Vitamin D Related/Nuclear Receptor, and Glucocorticoid Receptor
Catalog No. 50-003-48692