Learn More
Medchemexpress LLC Methylprednisolone-d2 | 83-43-2 | 99.1% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0260S41MG
Encompass_Preferred
Methylprednisolone-d2 is the deuterium labeled Methylprednisolone, a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. It improves severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05381