Learn More
Medchemexpress LLC Methylprednisolone-d2 | 83-43-2 | 99.1% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0260S45MG
Encompass_Preferred
Methylprednisolone-d2 is a deuterium-labeled synthetic corticosteroid known for its anti-inflammatory and immunomodulating effects. This compound has been shown to improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. It is also utilized as a tracer and an internal standard for various quantitative analyses, including NMR, GC-MS, and LC-MS.
  • Deuterium-labeled synthetic corticosteroid
  • Exhibits anti-inflammatory and immunomodulating properties
  • May improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels
  • Suitable for use as a tracer in quantitative analysis
  • Functions as an internal standard for NMR, GC-MS, or LC-MS
  • Deuteration may influence pharmacokinetic and metabolic profiles
Catalog No. 50-004-05860