Learn More
Medchemexpress LLC Methylprednisolone succinate | 2921-57-5 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1900100MG
Encompass_Preferred
Methylprednisolone succinate is a prodrug of Methylprednisolone and a glucocorticoid with immunosuppressant and anti-inflammatory activity. It binds to cytosolic glucocorticoid receptors, translocates to nuclei, and modulates target gene transcription. It alters Bax, Bcl-2, occludin, and ZO-1 expression, attenuates TLR4/NF-κB signaling, and suppresses proinflammatory cytokine production and immune cell activation. This product is for research use only.
  • Suppresses inflammatory responses
  • Attenuates blood-brain barrier disruption
  • Reduces neuronal apoptosis
  • Improves collateral blood flow into infarcted areas
  • Enhances metabolic response of ischemic myocardium
  • Modulates target gene transcription
  • Used for research on various inflammatory conditions and injuries
Catalog No. 50-004-11312