Learn More
Medchemexpress LLC Methylprednisolone succinate | 2921-57-5 | MFCD00271782 | 99.7% | 474.5 g·mol-1 | C26H34O8 | 10 MG

Supplier: Medchemexpress LLC HYB1900R10MG
Methylprednisolone succinate is a synthetic glucocorticoid research standard (prodrug of methylprednisolone) used as an anti-inflammatory reference in analytical and method-development applications. Supplied as a white to off-white solid for laboratory use; not for human or clinical use.
- Purity 99.73%.
- CAS number 2921-57-5.
- Molecular weight 474.5 g·mol-1.
- Available in small research sizes, including 10 mg.
- COA and SDS available; store per COA instructions.
- For research use only; not for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.