Learn More
Selleck Chemical LLC Methylproamine E8010-5MG
SDP
Supplier:  Selleck Chemical LLC E80105MG
Encompass_Preferred
Methylproamine is a DNA-binding radioprotector acts by repair of transient radiation-induced oxidative species on DNA Methylproamine also protects against ionizing radiation by preventing DNA double-strand breaks
Catalog No. 50-000-41655