Learn More
Medchemexpress LLC Methylstat | 1310877-95-2 | C28H31N3O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15221
Encompass_Preferred
Methylstat is a potent inhibitor of histone demethylases, showing anti-proliferative activity with low cytotoxicity. It promotes apoptosis and cell cycle arrest at the G0/G1 phase, while also increasing p53 and p21 protein expression. This compound inhibits angiogenesis induced by various cytokines, making it a valuable chemical probe for angiogenesis research.
  • Potent histone demethylases inhibitor
  • Exhibits anti-proliferative activity with low cytotoxicity
  • Induces apoptosis
  • Induces cell cycle arrest at the G0/G1 phase
  • Increases the expression of p53 and p21 protein levels
  • Inhibits angiogenesis induced by various cytokines
  • Can be utilized as a chemical probe to study its role in angiogenesis
Catalog No. 50-003-74583