Learn More
Cayman Chemical Methylstathydrate 10mg
SDP
Supplier:  Cayman Chemical 1109110 MG
Encompass_Preferred
A methyl ester prodrug of a JMJD inhibitor that has favorable cell permeability; inhibits growth of the JMJD2C-sensitive esophageal cancer cell line KYSE150 (GI50 = 5.1 µM); induces histone hypermethylation at multiple sites in a concentration-dependent manner
Catalog No. 50-295-5721