Learn More
Medchemexpress LLC Methyltetrazine-PEG4-acid | 1802907-91-0 | 99.8% | C20H28N4O7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14126450MG
Encompass_Preferred
Methyltetrazine-PEG4-acid is a PEG-based PROTAC linker designed for synthesizing PROTACs. It features methyltetrazine and carboxylic acid groups, enabling its use in copper-free click chemistry and reactions with primary amines.
  • Suitable for copper-free click chemistry
  • Reacts with primary amines
  • PEG-based linker structure
  • Utilized in PROTAC synthesis
  • Stable under recommended storage conditions
  • High purity (99.8%)
Catalog No. 50-003-53201