Learn More
Medchemexpress LLC Methyltetrazine-PEG4-amine hydrochloride | 2245150-18-7 | MFCD31813569 | 99.5% | C17H26ClN5O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY141261A10MG
Encompass_Preferred
Methyltetrazine-PEG4-amine hydrochloride is a PEG4-linked primary amine functionalized with a methyltetrazine group, provided as the hydrochloride salt. It functions as a click-chemistry reagent that undergoes rapid inverse-electron-demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) partners, enabling bioorthogonal labeling and site-specific conjugation in bioconjugation workflows. Supplied as a solid (purplish red to red) with high purity; store sealed at 4°C.
  • Enables rapid iEDDA click reactions with TCO partners.
  • PEG4 spacer improves solubility and flexibility.
  • Terminal primary amine allows straightforward conjugation chemistry.
  • Hydrochloride salt form enhances stability for storage.
  • High purity (~99.5%) suitable for labeling and conjugation.
  • Solid form, easy to handle and store at 4°C.
Catalog No. 50-002-59307