Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyltetrazine-PEG5-alkyne | 1802907-97-6 | 98.0% | C20H26N4O5 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY141275

Encompass_Preferred

Methyltetrazine-PEG5-alkyne is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It is provided as a pink to red solid with a purity of 98.0% for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Purity of 98.0%
  • Appearance: pink to red solid
  • Store at 4°C, under nitrogen
  • Soluble in DMSO

Catalog No. 50-003-52734


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.