Learn More
Medchemexpress LLC Methyltetrazine-PEG5-alkyne | 1802907-97-6 | MFCD28334550 | 98.0% | C20H26N4O5 | 10MG

Supplier: Medchemexpress LLC HY14127510MG
Methyltetrazine-PEG5-alkyne is a PEG-based linker reagent containing a methyltetrazine moiety and a terminal alkyne, intended for bioorthogonal conjugation and PROTAC linker synthesis. Supplied as a high-purity research chemical, it is suitable for click chemistry and bioconjugation workflows and is soluble in DMSO.
- Contains methyltetrazine and terminal alkyne functional groups.
- PEG5 spacer provides flexibility and solubility.
- High purity (98.0%) for reliable experimental results.
- Soluble in DMSO for easy handling and reaction setup.
- Available in small quantities suitable for research workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.