Learn More
Medchemexpress LLC Methyltetrazine-PEG5-NHS ester | 1802907-92-1 | MFCD28122951 | 97.0% | C24H31N5O9 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141269
Encompass_Preferred
Methyltetrazine-PEG5-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other to the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system for selective degradation of target proteins. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Solid appearance, purple to purplish red color
  • Solubility in DMSO: 100 mg/mL
  • Powder storage: -20°C for 3 years
  • For research use only
Catalog No. 50-003-52767