Learn More
Medchemexpress LLC Methyltetrazine-PEG8-PFP ester | 2353409-49-9 | MFCD31811432 | 98.1% | C34H43F5N4O11 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14127310MG
Encompass_Preferred
Methyltetrazine-PEG8-PFP ester is a PEG8-based linker featuring a methyltetrazine reactive group and a pentafluorophenyl (PFP) activated ester. Supplied as a powder for research use, it is intended for bioconjugation and PROTAC linker synthesis, enabling rapid tetrazine ligation and reliable amine coupling. Store protected from light at -20°C; in solution, store at -80°C for long-term stability.
Catalog No. 50-002-61299