Learn More
Apexbio Technology LLC Methylthioadenosine 2457-80-9 25mg
SDP
Supplier:  Apexbio Technology LLC B608025
Encompass_Preferred
Methylthioadenosine (2457-80-9) is a small-molecule inhibitor targeting high-affinity cyclic AMP phosphodiesterase It is designed to inhibit this enzyme thereby regulating cyclic AMP signaling and related cellular pathways Methylthioadenosine exerts its biological activity primarily through enzyme inhibition In cell-based studies it demonstrates inhibitory activity with a Ki value of 62 M against S49-derived high-affinity cyclic AMP phosphodiesterase Based on these pharmacological properties Methylthioadenosine holds research potential in exploring cellular signaling liver protection and mechanisms underlying hepatic pathologies
Catalog No. 50-000-92453