Learn More
Abcam 4-Methylumbelliferyl Beta-D-lactoside, 25MG
Supplier:  Abcam AB27389625MG
Encompass_Preferred
MW 500.4 Da, Purity >90%. Fluorogenic probe of ß-lactosidase or combined galactosidase and glucosidase activities. Releases the bright blue fluorophore 4-methylumbelliferone (4-MU) in direct correlation to the ß-lactosidase enzyme activity contained in the sample (EX 385 nm, EM 502 nm, e = 16K).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-124