Learn More
Apexbio Technology LLC Meticrane 1084-65-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A84715.1
Encompass_Preferred
Meticrane (CAS 1084-65-7) is a diuretic small molecule that acts on the distal convoluted tubules of the kidney inhibiting the reabsorption of sodium and chloride ions This action promotes increased urinary excretion contributing to the regulation of fluid and electrolyte homeostasis Meticrane is utilized in biomedical research focused on renal function ion transport mechanisms and is relevant for studies investigating the pathophysiology and pharmacology of primary hypertension
Catalog No. 50-000-95728