Learn More
Medchemexpress LLC Metipranolol hydrochloride | 36592-77-5 | 99.9% | 345.86 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16316100MG
Encompass_Preferred
Metipranolol hydrochloride is a non-selective β adrenergic receptor blocking agent. It is used systemically for the treatment of arterial hypertension and topically for the management of glaucoma.
  • Non-selective β adrenergic receptor blocking agent
  • For research use only
Catalog No. 50-003-94112