Learn More
Cayman Chemical MEtL314 Complexhumnrec 50ug
SDP
Supplier:  Cayman Chemical 2634250 UG
Encompass_Preferred
Source: Recombinant human N-terminal histidine-tagged METTL3 and recombinant human N-terminal GST-histidine-tagged METTL14 expressed in insect cells • Amino acids: 2-580 and 2-456 for METTL3 and METTL14, respectively • MW: 66.3 and 79.1 kDa for METTL3 and METTL14, respectively
Catalog No. 50-293-6421