Learn More
Medchemexpress LLC Metoclopramide | 364-62-5 | 99.9% | 299.80 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1738210MM/1ML
Encompass_Preferred
Metoclopramide is a potent antagonist of 5-HT3 and dopamine D2 receptors, with IC50s of 308 nM and 483 nM, respectively. It can be used for the research of nausea and vomiting, gastro-oesophageal reflux, and gastroparesis.
  • Potent antagonist of 5-HT3 and dopamine D2 receptor
  • Used for research of nausea and vomiting, gastro-oesophageal reflux, and gastroparesis
Catalog No. 50-003-84376