Learn More
Cayman Chemical MetoclopramIde-d3 500ug
SDP
Supplier:  Cayman Chemical 28885500 UG
Encompass_Preferred
An internal standard for the quantification of metoclopramide by GC- or LC-MS
Catalog No. 50-293-7484